missing translation for 'onlineSavingsMsg'
Learn More

Aconitine, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018899
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018899 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018899 Supplier Merck Emd Millipore Supplier No. 4478710MG
Food and Beverages

Specifications

CAS 302-27-2
Linear Formula C34H47 NO11
MDL Number MFCD00082375
Synonym Acetylbenzoylaconine
Type Analytical Standard
Quantity 10 mg
Formula Weight 645.74
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard