missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aconitase 1 Recombinant Rabbit Monoclonal Antibody (23GB3915)
GREENER_CHOICE

Catalog No. MA552636
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552636 100 μL
MA552637 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552636 Supplier Invitrogen™ Supplier No. MA552636
Only null left

Rabbit Recombinant Monoclonal Antibody

Aconitase 1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Aconitase 1 (ACO1) also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). It plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.

Specifications

Antigen Aconitase 1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3915
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACO1
Gene Accession No. P21399, P28271, Q63270
Gene Alias ACO1; Aco-1; Acon1; Aconitase; aconitase 1; aconitase 1, soluble; aconitate hydratase, cytoplasmic; ACONS; AH; Citrate hydro-lyase; cytoplasmic aconitase; cytoplasmic aconitate hydratase; cytosplasmic aconitase; epididymis luminal protein 60; ferritin repressor protein; FRP; HEL60; Ireb1; Irebp; IRE-BP 1; IREBP1; iron regulatory protein 1; iron-responsive element binding protein 1; iron-responsive element-binding protein 1; IRP1; OTTHUMP00000021176; OTTHUMP00000021177; OTTHUMP00000045233; soluble aconitase
Gene Symbols ACO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Aconitase 1 (15-42AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11428, 48, 50655
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less