missing translation for 'onlineSavingsMsg'
Learn More

Acid Yellow 23 98.0+%, TCI America™

Catalog No. F008825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F008825G 25 g
F0088500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F008825G Supplier TCI America Supplier No. F008825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1934-21-0
Molecular Formula C16H9N4Na3O9S2
Molecular Weight (g/mol) 534.356
MDL Number MFCD00148908
InChI Key UJMBCXLDXJUMFB-UHFFFAOYSA-K
Synonym tartrazine, yellow 5, aizen tartrazine, trisodium salt, food yellow 4, tartrazine fd&c yellow #5, c.i. acid yellow 23, atul tartrazine, erio tartrazine, kako tartrazine
PubChem CID 164825
IUPAC Name trisodium;5-oxo-1-(4-sulfonatophenyl)-4-[(4-sulfonatophenyl)diazenyl]-4H-pyrazole-3-carboxylate
SMILES C1=CC(=CC=C1N=NC2C(=NN(C2=O)C3=CC=C(C=C3)S(=O)(=O)[O-])C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 534.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acid Yellow 23
EINECSNumber (5)-1402
RTECSNumber UQ6400000
TSCA Yes