Learn More
Fisherbrand™ Acid Storage Standing Height Steel Cabinet
Fisherbrand™ Acid Storage Standing Height Steel Cabinets are used to store acids and are lined with a chemically resistant polyethylene liner.
Includes:
Vent Kit
Description
- Full height doors are lined with 1/8 in. thick polyethylene
- Chemical resistant 3/16 in. thick molded polyethylene lining with coved corners and 2 in. lip at front of cabinet opening
- Has one half-depth shelf
Compliance
SEFA 8
Specifications
| Acid Storage Cabinet | |
| 89.2 cm | |
| Polyethylene lining, Vent kit and one half-depth shelf | |
| 55.9 cm | |
| Standing Height Acid Storage Cabinet | |
| SEFA 8 | |
| Steel |
| 22 in. | |
| 35.1 in. | |
| 36 in. | |
| 91.4 cm | |
| 2 | |
| White |