missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 33, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014016
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014016 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014016 Supplier Merck Emd Millipore Supplier No. 5957325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 3567-66-6
Physical Form Neat
Linear Formula C16H11N3 Na2O7S2
MDL Number MFCD00071877
Synonym 5-Amino-4-hydroxy-3-phenylazo-2,7-naphthalenedisulfonic acid disodium salt; Acid Red 33 disodium salt; Disodium 5-amino-4-hydroxy-3-(phenylazo)-naphthalene-2,7-disulfonate; Food Red 12
Type Analytical Standard
Quantity 25 mg
Formula Weight 467.38
Percent Purity ≥97% (HPLC)
Grade Analytical Standard