missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 289, TCI America™

Catalog No. a190625g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A190625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A190625G Supplier TCI America Supplier No. A190625G
Only null left

Chemical Identifiers

CAS 12220-28-9
Molecular Formula C35H29N2NaO7S2
Molecular Weight (g/mol) 676.734
InChI Key MLYDRRZQNRGWEQ-UHFFFAOYSA-M
PubChem CID 92043221
IUPAC Name sodium;3-[[6-(2,6-dimethylphenyl)azaniumylidene-9-(2-sulfonatophenyl)xanthen-3-yl]amino]-2,4-dimethylbenzenesulfonate
SMILES CC1=C(C(=CC=C1)C)[NH+]=C2C=CC3=C(C4=C(C=C(C=C4)NC5=C(C=CC(=C5C)S(=O)(=O)[O-])C)OC3=C2)C6=CC=CC=C6S(=O)(=O)[O-].[Na+]

Specifications

Formula Weight 676.73
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Acid Red 289
EINECSNumber (5)-1577
TSCA Yes