accessibility menu, dialog, popup

UserName

CH3CH3HNHNH3CSOOOCH3OSOOONa(E/Z)(?)

CAS RN 12220-28-9

IUPAC Name: sodium 3-({3-[(2,6-dimethylphenyl)iminiumyl]-9-(2-sulfonatophenyl)-3H-xanthen-6-yl}amino)-2,4-dimethylbenzene-1-sulfonate
Synonyms: sodium 3-({6-[(2,6-dimethylphenyl)iminio]-9-(2-sulfonatophenyl)xanthen-3-yl}amino)-2,4-dimethylbenzenesulfonate
Molecular Weight (g/mol): 676.73
Molecular Formula: C35H29N2NaO7S2
InChi Key: MLYDRRZQNRGWEQ-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=CC(C)=C1[NH+]=C1C=CC2=C(C3=CC=C(NC4=C(C)C=CC(=C4C)S([O-])(=O)=O)C=C3OC2=C1)C1=CC=CC=C1S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results