missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Herbicide Mix in Methanol, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 11°C |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | Analytical Standard |