missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acid Fuchsin sodium salt

Catalog No. p-7026292
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2222214 25 g
AAB2222222 100 g
AAB2222236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2222214 Supplier Thermo Scientific Chemicals Supplier No. B2222214
Only null left

CAS: 3244-88-0 | C20H19N3Na2O9S3 | 587.544 g/mol

Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

Solubility
Soluble in water (1 mg/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3244-88-0
Molecular Formula C20H19N3Na2O9S3
Molecular Weight (g/mol) 587.544
MDL Number MFCD00013286
InChI Key VCEFFPMSOGDMJX-UHFFFAOYSA-N
Synonym acid fuchsin, acidal fuchsine, acid fuchsine, acidal magenta, fuchsine acid, fuchsin acid, p-fuchsine acid, acid fuchsine n, acid fuchsine o, acid fuchsine s
PubChem CID 131852436
IUPAC Name 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
SMILES CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]

Specifications

Quantity 25 g
Beilstein 4111656
Merck Index 14,107
Solubility Information Soluble in water (1mg/ml).
Formula Weight 585.53
Chemical Name or Material Acid Fuchsin sodium salt
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-816-5
RTECSNumber DD4737000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only