missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acid Fuchsin, pure, high purity biological stain
CAS: 3244-88-0 | C20H17N3Na2O9S3 | 587.544 g/mol
$150.40 - $437.07
Chemical Identifiers
| CAS | 3244-88-0 |
|---|---|
| Molecular Formula | C20H17N3Na2O9S3 |
| Molecular Weight (g/mol) | 587.544 |
| MDL Number | MFCD00013286 |
| InChI Key | VCEFFPMSOGDMJX-UHFFFAOYSA-N |
| Synonym | Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S |
| PubChem CID | 131852436 |
| IUPAC Name | 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium |
| SMILES | CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC227900250
|
Thermo Scientific Chemicals
227900250 |
25 g | Glass bottle |
Each for $150.40
|
|
||||
|
AC227901000
|
Thermo Scientific Chemicals
227901000 |
100 g | Glass bottle |
Each for $437.07
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3244-88-0 | |
| 587.544 | |
| VCEFFPMSOGDMJX-UHFFFAOYSA-N | |
| 131852436 | |
| CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] |
| C20H17N3Na2O9S3 | |
| MFCD00013286 | |
| Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S | |
| 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium |
Specifications
| 700 (at lambda min.) | |
| 1 to 1.4 | |
| MFCD00013286 | |
| Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol | |
| Authentic | |
| 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium | |
| 131852436 | |
| 585.52 | |
| Glass bottle | |
| 25 g | |
| Acid Fuchsin |
| 3244-88-0 | |
| C20H17N3Na2O9S3 | |
| Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S | |
| VCEFFPMSOGDMJX-UHFFFAOYSA-N | |
| CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na] | |
| 587.544 | |
| 544 to 550nm (in pH 3 acidified water) | |
| Pure | |
| Green | |
| Powder or Crystals |
Safety and Handling
EINECSNumber : 221-816-5
RTECSNumber : DD4737000
TSCA : TSCA
RUO – Research Use Only