missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acid Fuchsin, pure, high purity biological stain

CAS: 3244-88-0 | C20H17N3Na2O9S3 | 587.544 g/mol

Supplier:  Thermo Scientific Chemicals 227901000

 View more versions of this product

Catalog No. AC227901000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

700 (at lambda min.)
3244-88-0
C20H17N3Na2O9S3
Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S
VCEFFPMSOGDMJX-UHFFFAOYSA-N
CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]
587.544
544 to 550nm (in pH 3 acidified water)
Pure
Green
Powder or Crystals
Acid Fuchsin
1 to 1.4
MFCD00013286
Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol
Authentic
3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
131852436
585.52
Glass bottle
100 g

Chemical Identifiers

3244-88-0
587.544
VCEFFPMSOGDMJX-UHFFFAOYSA-N
131852436
CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]
C20H17N3Na2O9S3
MFCD00013286
Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S
3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-816-5

RTECSNumber : DD4737000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only