missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Digestion Reagent (with Mercury), For Organic Nitrogen, LabChem™
Supplier: LabChem LC105502
Specifications
| Acid Digestion Reagent (with Mercury) | |
| For Organic Nitrogen | |
| 7664-93-9,7778-80-5,7783-35-9 | |
| UN2796 | |
| Soluble in water | |
| Passes Test | |
| 1.2g/mL | |
| 1 L |
| Colorless | |
| 56.9,29.5,13.4,0.2 | |
| HgSO4 | |
| >1.2g/mL | |
| 296.65 | |
| Poly Bottle | |
| Sulfur compounds; Mercury; Thermal decomposition generates: Corrosive vapors | |
| Liquid |