missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Digestion Reagent, For Organic Nitrogen, LabChem™
Supplier: LabChem LC105602
Specifications
| Acid Digestion Reagent | |
| For Organic Nitrogen | |
| 7778-80-5,7664-93-9,7758-99-8 | |
| UN2796 | |
| 249.68 | |
| Poly Bottle | |
| 1 L |
| Blue | |
| 72.06,13.4,13.4,1.14 | |
| CuSO4·5H2O | |
| Soluble in water | |
| Passes Test | |
| Sulfur compounds; Copper; Thermal decomposition generates: Corrosive vapors | |
| Liquid |