Learn More
Parr™ Acid Digestion Bomb Replacement Cups
For use with Parr General Purpose Acid Digestion Bombs
Supplier: Parr™ A280AC2
Specifications
| Cup with Cover | |
| 23 mL | |
| 4744 general purpose acid digestion bomb |
| 0.77 oz. | |
| 4744 general purpose acid digestion bomb | |
| PTFE |