missing translation for 'onlineSavingsMsg'
Learn More

Acid Compounds - WP, MilliporeSigma™ Supelco™

Catalog No. 111017059
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017059 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017059 Supplier Merck Emd Millipore Supplier No. PE12741.2ML

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 67-64-1
Flash Point -17°C - closed cup - Solvent
Type Proficiency Testing Material
Quantity 1.2 mL