missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Blue 29, MP Biomedicals™
$74.51 - $197.83
Chemical Identifiers
| CAS | 5850-35-1 |
|---|---|
| Molecular Formula | C22H14N6Na2O9S2 |
| Molecular Weight (g/mol) | 616.49 |
| MDL Number | MFCD00009960 |
| InChI Key | WEILTBATXPPXPP-AXSRUCDFSA-L |
| Synonym | C.I. 20460 |
| PubChem CID | 44134491 |
| IUPAC Name | disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate |
| SMILES | [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15259625
|
MP Biomedicals Inc
0215259625 |
25 g |
Each for $74.51
|
|
|||||
|
ICN15259680
|
MP Biomedicals Inc
0215259680 |
100 g |
Each for $197.83
|
|
|||||
Description
Stains and dyesChemical Identifiers
| 5850-35-1 | |
| 616.49 | |
| WEILTBATXPPXPP-AXSRUCDFSA-L | |
| 44134491 | |
| [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O |
| C22H14N6Na2O9S2 | |
| MFCD00009960 | |
| C.I. 20460 | |
| disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate |
Specifications
| 5850-35-1 | |
| MFCD00009960 | |
| WEILTBATXPPXPP-AXSRUCDFSA-L | |
| disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate | |
| 44134491 | |
| Black | |
| Powder |
| C22H14N6Na2O9S2 | |
| C.I. 20460 | |
| [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O | |
| 616.49 | |
| 616.5 | |
| 25 g | |
| Acid Blue 29 |
Safety and Handling
EINECSNumber : 227-449-7
Recommended Storage : Store at Room Temperature (15° to 30°C)