accessibility menu, dialog, popup

UserName

NaNaNH2NNNOONHNOSOOOSOOO(E/Z)(E)

CAS RN 5850-35-1

IUPAC Name: disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (6E)-4-amino-3-[2-(3-nitrophenyl)diazen-1-yl]-5-oxo-6-(2-phenylhydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 616.49
Molecular Formula: C22H14N6Na2O9S2
InChi Key: WEILTBATXPPXPP-AXSRUCDFSA-L
SMILES: [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)