missing translation for 'onlineSavingsMsg'
Learn More

Acid Blue 29, MP Biomedicals™

Catalog No. ICN15259625
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15259625 25 g
ICN15259680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15259625 Supplier MP Biomedicals Inc Supplier No. 0215259625
Only null left
Stains and dyes

Chemical Identifiers

CAS 5850-35-1
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
MDL Number MFCD00009960
InChI Key WEILTBATXPPXPP-AXSRUCDFSA-L
Synonym C.I. 20460
PubChem CID 44134491
IUPAC Name disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Formula Weight 616.5
Color Black
Quantity 25 g
Physical Form Powder
Chemical Name or Material Acid Blue 29
EINECSNumber 227-449-7
Recommended Storage Store at Room Temperature (15° to 30°C)