Learn More
Acid-Alcohol, 5% (v/v) in 95% Alcohol, Certified, 5.0% (v/v) ±0.2%, LabChem™
$119.03 - $119.03
Chemical Identifiers
| CAS | 64-17-5 |
|---|---|
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| PubChem CID | 702 |
| ChEBI | CHEBI:16236 |
Chemical Identifiers
| 64-17-5 | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| 702 | |
| CHEBI:16236 |
Specifications
| Liquid | |
| 86.5,1.5,4.5,3.5,4 | |
| Soluble in water | |
| Colorless | |
| HCl | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| CHEBI:16236 | |
| 5.0% ±0.2% (v/v) | |
| Acid-Alcohol, 5% (v/v) in 95% Alcohol |
| 64-17-5,7647-01-0,67-63-0,67-56-1,7732-18-5 | |
| Poly Bottle | |
| Hydrogen chloride; Carbon monoxide; Carbon dioxide; May release flammable gases; Thermal decomposition generates: Corrosive vapors | |
| 1 L | |
| UN2924 | |
| 702 | |
| 36.46 | |
| Certified |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature