missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Acetylthio)acetic Acid 98.0+%, TCI America™
$201.32 - $767.98
Chemical Identifiers
| CAS | 1190-93-8 |
|---|---|
| Molecular Formula | C4H6O3S |
| Molecular Weight (g/mol) | 134.149 |
| MDL Number | MFCD00021763 |
| InChI Key | QSBWDKUBOZHGOU-UHFFFAOYSA-N |
| PubChem CID | 14485 |
| IUPAC Name | 2-acetylsulfanylacetic acid |
| SMILES | CC(=O)SCC(=O)O |
Chemical Identifiers
| 1190-93-8 | |
| 134.149 | |
| QSBWDKUBOZHGOU-UHFFFAOYSA-N | |
| 2-acetylsulfanylacetic acid |
| C4H6O3S | |
| MFCD00021763 | |
| 14485 | |
| CC(=O)SCC(=O)O |
Specifications
| 1190-93-8 | |
| 160°C | |
| MFCD00021763 | |
| QSBWDKUBOZHGOU-UHFFFAOYSA-N | |
| 2-acetylsulfanylacetic acid | |
| 14485 | |
| ≥98.0% (GC,T) | |
| (Acetylthio)acetic Acid |
| Yellow | |
| C4H6O3S | |
| 5 g | |
| CC(=O)SCC(=O)O | |
| 134.149 | |
| 134.15 | |
| Liquid |
Safety and Handling
RTECSNumber : AF1665000
TSCA : Yes