accessibility menu, dialog, popup

UserName

H3COSOHO

CAS RN 1190-93-8

IUPAC Name: 2-(acetylsulfanyl)acetic acid
Synonyms: acetic acid, (acetylthio)-
Molecular Weight (g/mol): 134.15
Molecular Formula: C4H6O3S
InChi Key: QSBWDKUBOZHGOU-UHFFFAOYSA-N
SMILES: CC(=O)SCC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results