missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Acetylthio)acetic Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 160°C |
| Quantity | 5 g |
| Formula Weight | 134.15 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | (Acetylthio)acetic Acid |
Safety and Handling
| RTECSNumber | AF1665000 |
| TSCA | Yes |