missing translation for 'onlineSavingsMsg'
Learn More

Acetylacetone 99.0+%, TCI America™

Catalog No. p-7134396
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P005225ML 25 mL
P0052500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P005225ML Supplier TCI America Supplier No. P005225ML
Only null left

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O

Specifications

Melting Point -23°C
Color Yellow
Boiling Point 138°C
Quantity 25 mL
UN Number 2310
Formula Weight 100.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetylacetone
EINECSNumber (2)-0562
RTECSNumber SA1925000
TSCA Yes