missing translation for 'onlineSavingsMsg'
Learn More

Acetylacetonatobis(ethylene)rhodium(I), TCI America™

Catalog No. A2100200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2100200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2100200MG Supplier TCI America Supplier No. A2100200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12082-47-2
Molecular Formula C9H15O2Rh
Molecular Weight (g/mol) 258.12
MDL Number MFCD00015354
InChI Key FLRBEQQDEGBCJS-FGSKAQBVSA-M
Synonym acetylacetonatobis ethylene rhodium i, bis ethylene 2,4-pentanedionato rhodium i, acetylacetonatobis ethylene rhodium, ethylene; z-4-oxopent-2-en-2-olate; rhodium
PubChem CID 11630270
IUPAC Name λ¹-rhodium(1+) bis(ethene) (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].C=C.C=C.C\C([O-])=C\C(C)=O

Specifications

Melting Point 141°C
Quantity 200 mg
Formula Weight 258.12
Physical Form Crystalline Powder
Chemical Name or Material Acetylacetonatobis(ethylene)rhodium(I)
TSCA No
Recommended Storage Refrigerator