accessibility menu, dialog, popup

UserName

OOH3CH3CRh

CAS RN 12082-47-2

IUPAC Name: λ¹-rhodium(1+) bis(ethene) (2Z)-4-oxopent-2-en-2-olate
Synonyms: λ¹-rhodium(1+) bis(ethylene) (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 258.12
Molecular Formula: C9H15O2Rh
InChi Key: FLRBEQQDEGBCJS-FGSKAQBVSA-M
SMILES: [Rh+].C=C.C=C.C\C([O-])=C\C(C)=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)