missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Acetylacetonato)(1,5-cyclooctadiene)iridium(I), TCI America™
$227.41 - $680.38
Chemical Identifiers
| CAS | 12154-84-6 |
|---|---|
| Molecular Formula | C13H19IrO2 |
| Molecular Weight (g/mol) | 399.51 |
| MDL Number | MFCD08273779 |
| InChI Key | RYGIGGDIKNVIPI-DWVXZKBMSA-M |
| Synonym | Ir(acac)(COD) |
| IUPAC Name | iridium(2+) (1Z,5Z)-cycloocta-1,5-diene (2Z)-4-oxopent-2-en-2-olate |
| SMILES | [Ir++].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2981200MG
|
TCI America
A2981200MG |
200 mg |
Each for $227.41
|
|
|||||
|
A29811G
|
TCI America
A29811G |
1 g |
Each for $680.38
|
|
|||||
Chemical Identifiers
| 12154-84-6 | |
| 399.51 | |
| RYGIGGDIKNVIPI-DWVXZKBMSA-M | |
| iridium(2+) (1Z,5Z)-cycloocta-1,5-diene (2Z)-4-oxopent-2-en-2-olate |
| C13H19IrO2 | |
| MFCD08273779 | |
| Ir(acac)(COD) | |
| [Ir++].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1 |
Specifications
| 12154-84-6 | |
| C13H19IrO2 | |
| 200 mg | |
| RYGIGGDIKNVIPI-DWVXZKBMSA-M | |
| iridium(2+) (1Z,5Z)-cycloocta-1,5-diene (2Z)-4-oxopent-2-en-2-olate | |
| 399.51 | |
| (Acetylacetonato)(1,5-cyclooctadiene)iridium(I) |
| 155°C | |
| MFCD08273779 | |
| Ir(acac)(COD) | |
| [Ir++].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1 | |
| 399.51 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NO3615000
TSCA : No