accessibility menu, dialog, popup

UserName

CAS RN 12154-84-6

IrOOCH3CH3

CAS RN 12154-84-6

IUPAC Name: iridium(2+) (1Z,5Z)-cycloocta-1,5-diene (2Z)-4-oxopent-2-en-2-olate
Synonyms: iridium(2+) 1,5-cyclooctadiene, (Z,Z)- (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 399.51
Molecular Formula: C13H19IrO2
InChi Key: RYGIGGDIKNVIPI-DWVXZKBMSA-M
SMILES: [Ir++].C\C([O-])=C\C(C)=O.C1C\C=C/CC\C=C/1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results