missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Acetylacetonato)(1,5-cyclooctadiene)iridium(I), TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Quantity | 200 mg |
| Formula Weight | 399.51 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (Acetylacetonato)(1,5-cyclooctadiene)iridium(I) |
Safety and Handling
| RTECSNumber | NO3615000 |
| TSCA | No |