missing translation for 'onlineSavingsMsg'
Learn More

Medisca Acetyl-L-Carnitine Hydrochloride (Dietary Supplement Grade)

Catalog No. 14186235
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186235 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14186235 Supplier Medisca Supplier No. 095805
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetyl-L-Carnitine Hydrochloride (Dietary Supplement Grade) is a white to off-white, highly hygroscopic crystalline powder with a vinegar-like odor.

Acetyl-L-Carnitine Hydrochloride is a white to off-white, highly hygroscopic crystalline powder with a vinegar-like odor.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Acetyl levocarnitine hydrochloride, Acetyl L-carnitine, Acetylcarnitine chloride, L-Acetylcarnitine Hydrochloride

Specifications

Chemical Name or Material Acetyl-L-Carnitine Hydrochloride (Dietary Supplement Grade)
Packaging Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck
CAS 5080-50-2
Type Drug Standard
Molecular Weight (g/mol) 239.6972
Quantity 100 g
Percent Purity 98 to 101 %
Grade Ungraded