missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210049080
Specifications
| Acetyl Coenzyme A Trilithium Salt Trihydrate | |
| White | |
| C23H38Li3N7O17P3S | |
| acetyl coenzyme a trilithium salt | |
| [Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O | |
| 830.39 | |
| 881.1 |
| 75520-41-1 | |
| >92% | |
| 100 mg | |
| MOGXDJCKYPGZRO-JHJDYNLLSA-N | |
| S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium | |
| 129893900 | |
| Powder |
Chemical Identifiers
| 75520-41-1 | |
| 830.39 | |
| acetyl coenzyme a trilithium salt | |
| S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium |
| C23H38Li3N7O17P3S | |
| MOGXDJCKYPGZRO-JHJDYNLLSA-N | |
| 129893900 | |
| [Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O |
Safety and Handling
Recommended Storage : Store at -20°C.