missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™

Catalog No. p-4589721
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10049001 1 mg
ICN10049005 5 mg
ICN10049010 10 mg
ICN10049025 25 mg
ICN10049080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10049001 Supplier MP Biomedicals Inc Supplier No. 0210049001
Only null left

Chemical Identifiers

CAS 75520-41-1
Molecular Formula C23H38Li3N7O17P3S
Molecular Weight (g/mol) 830.39
InChI Key MOGXDJCKYPGZRO-JHJDYNLLSA-N
Synonym acetyl coenzyme a trilithium salt
PubChem CID 129893900
IUPAC Name S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium
SMILES [Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O

Specifications

Color White
Assay Percent Range >92%
Quantity 1 mg
Formula Weight 881.1
Physical Form Powder
Chemical Name or Material Acetyl Coenzyme A Trilithium Salt Trihydrate
Recommended Storage Store at -20°C.