missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone p-Toluenesulfonylhydrazone 99.0+%, TCI America™

Catalog No. A21745G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21745G Supplier TCI America Supplier No. A21745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4545-21-5
Molecular Formula C15H16N2O2S
Molecular Weight (g/mol) 288.365
MDL Number MFCD01140191
InChI Key MIXFDFCKBMCLGN-SSZFMOIBSA-N
Synonym acetophenone tosylhydrazone, 4-methyl-n'-1z-1-phenylethylidene benzenesulfonohydrazide, acetophenonetosylhydrazone, 4-methyl-n'-1e-1-phenylethylidene benzenesulfonohydrazide, acetophenone tosyl hydrazone, 4-methyl-n-z-1-phenylethylideneamino benzenesulfonamide
PubChem CID 6287802
IUPAC Name 4-methyl-N-[(Z)-1-phenylethylideneamino]benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=C(C)C2=CC=CC=C2

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 5 g
Formula Weight 288.37
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetophenone p-Toluenesulfonylhydrazone