missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone p-toluenesulfonylhydrazone, 98%

Catalog No. p-7035747
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5347406 5 g
AAH5347414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5347406 Supplier Thermo Scientific Chemicals Supplier No. H5347406
Only null left

CAS: 4545-21-5 | C15H16N2O2S | 288.365 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4545-21-5
Molecular Formula C15H16N2O2S
Molecular Weight (g/mol) 288.365
MDL Number MFCD01140191
InChI Key MIXFDFCKBMCLGN-SSZFMOIBSA-N
Synonym acetophenone tosylhydrazone, 4-methyl-n'-1z-1-phenylethylidene benzenesulfonohydrazide, acetophenonetosylhydrazone, 4-methyl-n'-1e-1-phenylethylidene benzenesulfonohydrazide, acetophenone tosyl hydrazone, 4-methyl-n-z-1-phenylethylideneamino benzenesulfonamide
PubChem CID 6287802
IUPAC Name 4-methyl-N-[(Z)-1-phenylethylideneamino]benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN=C(C)C2=CC=CC=C2

Specifications

Melting Point 144°C to 149°C
Quantity 5 g
Formula Weight 288.36
Percent Purity 98%
Chemical Name or Material Acetophenone p-toluenesulfonylhydrazone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only