missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone Oxime 98.0+%, TCI America™

Catalog No. A080725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A080725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A080725G Supplier TCI America Supplier No. A080725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 613-91-2
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00013931 MFCD00013931
InChI Key JHNRZXQVBKRYKN-VQHVLOKHSA-N
Synonym acetophenone oxime, ethanone, 1-phenyl-, oxime, acetophenone, oxime, methyl phenyl ketone oxime, methyl phenyl ketoxime, 1-phenylethanone oxime, z-acetophenone oxime, n-1-phenylethylidene hydroxylamine, e-n-1-phenylethylidene hydroxylamine, ne-n-1-phenylethylidene hydroxylamine
PubChem CID 5464950
IUPAC Name (E)-N-(1-phenylethylidene)hydroxylamine
SMILES C\C(=N/O)C1=CC=CC=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 119°C
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Acetophenone Oxime
RTECSNumber AM9654000
TSCA Yes