accessibility menu, dialog, popup

UserName

CAS RN 613-91-2

IUPAC Name: (E)-N-(1-phenylethylidene)hydroxylamine
Synonyms: ethanone, 1-phenyl-, oxime
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: JHNRZXQVBKRYKN-VQHVLOKHSA-N
SMILES: C\C(=N/O)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)