Learn More
Acetophenone oxime, 98%
CAS: 613-91-2 | C8H9NO | 135.17 g/mol
Supplier: Thermo Scientific Chemicals A1180418
Description
Used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.
Solubility
Slightly soluble in water. Soluble in ethyl alcohol almost transparency.
Notes
Air and Moisture Sensitive. Store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| Acetophenone oxime | |
| 58°C to 60°C | |
| C8H9NO | |
| 50 g | |
| Air and moisture sensitive | |
| Slightly soluble in water. Soluble in ethyl alcohol almost transparency. | |
| C\C(=N/O)C1=CC=CC=C1 | |
| 135.17 | |
| 135.16 |
| 613-91-2 | |
| 118°C to 120°C (20 mmHg) | |
| MFCD00013931 MFCD00013931 | |
| 1562059 | |
| acetophenone oxime, ethanone, 1-phenyl-, oxime, acetophenone, oxime, methyl phenyl ketone oxime, methyl phenyl ketoxime, 1-phenylethanone oxime, z-acetophenone oxime, n-1-phenylethylidene hydroxylamine, e-n-1-phenylethylidene hydroxylamine, ne-n-1-phenylethylidene hydroxylamine | |
| JHNRZXQVBKRYKN-VQHVLOKHSA-N | |
| (NE)-N-(1-phenylethylidene)hydroxylamine | |
| 5464950 | |
| 98% |
Chemical Identifiers
| 613-91-2 | |
| 135.17 | |
| JHNRZXQVBKRYKN-VQHVLOKHSA-N | |
| 5464950 | |
| C\C(=N/O)C1=CC=CC=C1 |
| C8H9NO | |
| MFCD00013931 MFCD00013931 | |
| acetophenone oxime, ethanone, 1-phenyl-, oxime, acetophenone, oxime, methyl phenyl ketone oxime, methyl phenyl ketoxime, 1-phenylethanone oxime, z-acetophenone oxime, n-1-phenylethylidene hydroxylamine, e-n-1-phenylethylidene hydroxylamine, ne-n-1-phenylethylidene hydroxylamine | |
| (NE)-N-(1-phenylethylidene)hydroxylamine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-360-2
RTECSNumber : AM9654000
TSCA : Yes
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only