missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone azine, 97%

Catalog No. aal0392406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0392406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0392406 Supplier Thermo Scientific Chemicals Supplier No. L0392406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 729-43-1 | C16H16N2 | 236.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 729-43-1
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.318
MDL Number MFCD00015011
InChI Key MOKMQSIJAHPSQX-SIJTWYJSSA-N
Synonym acetophenone azine, 1-phenyl-n-z-1-phenylethylideneamino ethanimine
PubChem CID 5914692
IUPAC Name (E)-1-phenyl-N-[(Z)-1-phenylethylideneamino]ethanimine
SMILES CC(=NN=C(C)C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 120°C to 124°C
Quantity 5 g
Beilstein 1912471
Formula Weight 236.32
Percent Purity 97%
Chemical Name or Material Acetophenone azine
EINECSNumber 211-979-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only