accessibility menu, dialog, popup

UserName

CAS RN 729-43-1

IUPAC Name: (E,E)-bis(1-phenylethylidene)hydrazine
Synonyms: (E,E)-bis(1-phenylethylidene)hydrazine
Molecular Weight (g/mol): 236.32
Molecular Formula: C16H16N2
InChi Key: MOKMQSIJAHPSQX-HBKJEHTGSA-N
SMILES: C\C(=N/N=C(\C)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results