missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile:Water 55:45 with 0.32% Acetic Acid, Reagents

Catalog No. 01335927
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335927 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335927 Supplier Labchem Inc Supplier No. CA1078004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetonitrile:Water 55:45 with 0.32% Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 75-05-8 , 75-05-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methyl cyanide
IUPAC Name water
SMILES O

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8844
Boiling Point 76°C
Percent Purity 55%
Grade Reagent
Chemical Name or Material Acetonitrile:Water 55:45 with 0.32% Acetic Acid