missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023039
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023039 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023039 Supplier Merck Emd Millipore Supplier No. RTC00008020ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 75-05-8
Vapor Pressure 72.8 mmHg (20°C)
Vapor Density 1.41 (vs air)
Density 0.786 g/mL (at 25°C (literature))
Flash Point 2°C (Not Applicable)
Molecular Formula C2H3N
Linear Formula CH3CN
MDL Number MFCD00001878
Melting Point -45°C (literature)
Boiling Point 81°C to 82°C (literature)
Synonym Acetonitrile; ACN; Cyanomethane; Ethyl nitrile; Methyl cyanide
Quantity 20 mL
Formula Weight 41.05
Grade Certified Reference Material
Show More Show Less