missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Technical, Reagents

Catalog No. p-200021216
Change view
Click to view available options
Quantity:
4 L
20 L
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337612 20 L
01337611 4 L
01337610 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337612 Supplier Labchem Inc Supplier No. C210260020E6
Only null left

Acetone, Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.788
Boiling Point 56°C
Grade Technical
Chemical Name or Material Acetone, Technical