Learn More
Acetone, specified according to the requirements of Ph.Eur.
CAS: 67-64-1 | C3H6O | 58.08 g/mol
$90.60 - $90.60
Chemical Identifiers
| CAS | 67-64-1 |
|---|---|
| Molecular Formula | C3H6O |
| Molecular Weight (g/mol) | 58.08 |
| MDL Number | MFCD00008765 |
| InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
| IUPAC Name | propan-2-one |
| SMILES | CC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC447450010
|
Thermo Scientific Chemicals
447450010 |
1 L |
Each for $90.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| CC(C)=O |
| C3H6O | |
| MFCD00008765 | |
| propan-2-one |
Specifications
| -95.0°C | |
| 0.7900 g/mL | |
| 1 L | |
| Glass Bottle | |
| 0.79 | |
| CH3COCH3 | |
| 0.32 mPa.s (20°C) | |
| Solubility aq. soln.: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils | |
| CC(C)=O | |
| 58.08 | |
| Acetone |
| 67-64-1 | |
| 56.0°C | |
| -20°C | |
| 1.3585 | |
| C3H6O | |
| MFCD00008765 | |
| 15, 65 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 58.08 |
Safety and Handling
GHS H Statement May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Ground and bond container and receiving equipment
GHS Signal Word: Danger
EINECSNumber : 200-662-2
RUO – Research Use Only