missing translation for 'onlineSavingsMsg'
Learn More

Acetone, specified according to the requirements of Ph.Eur.

Catalog No. AC447450010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447450010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC447450010 Supplier Thermo Scientific Chemicals Supplier No. 447450010

CAS: 67-64-1 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
Density 0.7900 g/mL
Quantity 1 L
Flash Point -20°C
Packaging Glass Bottle
Refractive Index 1.3585
Specific Gravity 0.79
Linear Formula CH3COCH3
Viscosity 0.32 mPa.s (20°C)
Merck Index 15, 65
Boiling Point 56.0°C
Solubility Information Solubility aq. soln.: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Chemical Name or Material Acetone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause drowsiness or dizziness.

Causes serious eye irritation.

Highly flammable liquid and vapor.

Repeated exposure may cause skin dryness or cracking.

Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Ground and bond container and receiving equipment
EINECSNumber 200-662-2

RUO – Research Use Only