missing translation for 'onlineSavingsMsg'
Learn More

Acetone Semicarbazone 98.0+%, TCI America™

Supplier:  TCI America A0058500G

 View more versions of this product

Catalog No. A0058500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetone Semicarbazone
187°C
C4H9N3O
500 g
HQDAJGNZGNZGCO-UHFFFAOYSA-N
(propan-2-ylideneamino)urea
66965
≥98.0% (T)
110-20-3
White-Yellow
MFCD00014785
acetone semicarbazone, acetonesemicarbazone, acetone, semicarbazone, 2-propanone, semicarbazone, hydrazinecarboxamide, 2-1-methylethylidene, isopropylideneamino urea, propan-2-ylideneamino urea, 2-propan-2-ylidene hydrazinecarboxamide, 2-propanone semicarbazone, acmc-1bs9q
CC(=NNC(=O)N)C
115.136
115.14
Crystalline Powder

Chemical Identifiers

110-20-3
115.136
HQDAJGNZGNZGCO-UHFFFAOYSA-N
66965
CC(=NNC(=O)N)C
C4H9N3O
MFCD00014785
acetone semicarbazone, acetonesemicarbazone, acetone, semicarbazone, 2-propanone, semicarbazone, hydrazinecarboxamide, 2-1-methylethylidene, isopropylideneamino urea, propan-2-ylideneamino urea, 2-propan-2-ylidene hydrazinecarboxamide, 2-propanone semicarbazone, acmc-1bs9q
(propan-2-ylideneamino)urea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0041

RTECSNumber : AL7175000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions