accessibility menu, dialog, popup

UserName

CAS RN 110-20-3

IUPAC Name: [(propan-2-ylidene)amino]urea
Synonyms: acetone, semicarbazone
Molecular Weight (g/mol): 115.14
Molecular Formula: C4H9N3O
InChi Key: HQDAJGNZGNZGCO-UHFFFAOYSA-N
SMILES: CC(C)=NNC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results