missing translation for 'onlineSavingsMsg'
Learn More

Acetone Semicarbazone 98.0+%, TCI America™

Catalog No. A005825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A005825G 25 g
A0058500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A005825G Supplier TCI America Supplier No. A005825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-20-3
Molecular Formula C4H9N3O
Molecular Weight (g/mol) 115.136
MDL Number MFCD00014785
InChI Key HQDAJGNZGNZGCO-UHFFFAOYSA-N
Synonym acetone semicarbazone, acetonesemicarbazone, acetone, semicarbazone, 2-propanone, semicarbazone, hydrazinecarboxamide, 2-1-methylethylidene, isopropylideneamino urea, propan-2-ylideneamino urea, 2-propan-2-ylidene hydrazinecarboxamide, 2-propanone semicarbazone, acmc-1bs9q
PubChem CID 66965
IUPAC Name (propan-2-ylideneamino)urea
SMILES CC(=NNC(=O)N)C

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 25 g
Formula Weight 115.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetone Semicarbazone
EINECSNumber (9)-0041
RTECSNumber AL7175000
TSCA Yes