Learn More
Acetone, for residue analysis, Thermo Scientific™
Chemical Identifiers
| CAS | 67-64-1 |
|---|---|
| Molecular Formula | C3H6O |
| Molecular Weight (g/mol) | 58.08 |
| MDL Number | MFCD00008765 |
| InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
| Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
| PubChem CID | 180 |
| ChEBI | CHEBI:15347 |
| IUPAC Name | propan-2-one |
| SMILES | CC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC326570010
|
Thermo Scientific Chemicals
326570010 |
1L | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC326570025
|
Thermo Scientific Chemicals
326570025 |
2.5L | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For residue analysisChemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| 180 | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CHEBI:15347 | |
| CC(C)=O |
Specifications
| 67-64-1 | |
| 56.0°C | |
| −20°C | |
| 1.3585 | |
| 0.79 | |
| CH3COCH3 | |
| 0.32 mPa.s (20°C) | |
| 01, 635 | |
| 15, 65 | |
| Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| Residue Analysis |
| 0.7900g/mL | |
| 1L | |
| Glass bottle | |
| 2ppm max. | |
| C3H6O | |
| MFCD00008765 | |
| 0.2% max. (coulometric) | |
| 02,13; 03,4; 06,9 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| 58.08 | |
| Acetone, ECD Tested for Pesticide Analysis |
Safety and Handling
GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 200-662-2
RTECSNumber : AL3150000
TSCA : TSCA
RUO â Research Use Only