missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acétone, pour l’analyse des résidus
Description
Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.
Pour l’analyse des résidus
Spécifications
Spécifications
| Melting Point | -95.0°C |
| Density | 0.7900g/mL |
| Quantity | 2.5 L |
| Flash Point | −20°C |
| Packaging | Bouteille en verre |
| Physical Form | Liquide |
| Refractive Index | 1.3585 |
| Residue after Evaporation | 2ppm max. |
| Specific Gravity | 0.79 |
| Linear Formula | CH3COCH3 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause drowsiness or dizziness. Causes serious eye irritation. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 200-662-2 |
| RTECSNumber | AL3150000 |
| TSCA | TSCA |
Utilisation uniquement de la recherche RUO –