missing translation for 'onlineSavingsMsg'
Learn More

Acetone, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.5% (GC), Honeywell™ Riedel-de Haën™

Catalog No. 60046066
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Poly Bottle
Stainless Steel Can
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046066 20 kg Stainless Steel Can
60046062 1 L Poly Bottle
60046064 2.5 L Glass Bottle
60046063 2.5 L Poly Bottle
60046070 6 x 1 L Poly Bottle
60046069 4 x 2.5 L Glass Bottle
60046068 4 x 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60046066 Supplier Honeywell Inc Supplier No. 3220120KG
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C
Appearance Colorless liquid
Density 0.791g/mL at 25°C
Quantity 20 kg
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Stainless Steel Can
Refractive Index n20/D 1.359
Residue after Evaporation 0 to 0.001%
Titratable Acid ≤0.0015% (as CH3COOH)
Titratable Base ≤0.0008% (as NH3)
Vapor Pressure 184mmHg (20°C)
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Boiling Point 56°C (760 mmHg)
Formula Weight 58.08g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Acetone, Reag. Ph. Eur., reag. ISO
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Acetone
EINECSNumber 200-662-2
RTECSNumber AL3150000