missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™

Catalog No. 60045553
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045553 1 L Poly Bottle
60045555 2.5 L Glass Bottle
60045554 2.5 L Poly Bottle
60045561 5 L Poly Bottle
60045562 6 x 1 L Poly Bottle
60045559 4 x 2.5 L Glass Bottle
60045558 4 x 2.5 L Poly Bottle
60045556 20 kg Stainless Steel Can
60045560 4 x 5 L Poly Bottle
60045552 155 kg Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 60045553 Supplier Honeywell Inc Supplier No. 242011L
Only null left

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95°C
Color Colorless
Density 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 184mmHg (20°C)
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Boiling Point 55°C to 57°C
Formula Weight 58.08g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Acetone, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Acetone
EINECSNumber 200-662-2
RTECSNumber AL3150000